(3,4-Diethoxyphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
2-(3,4-Diethoxyphenyl)acetonitrile
018552-1G
FLUO018552-1GEA
162
EUR
FLUO018552-1G
(3,4-Diethoxyphenyl)acetonitril
(3,4-Diethoxyphenyl)acetonitril
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205,253 g/mol |
MDL:
MFCD00060715 CAS-Nummer: 27472-21-5 EINECS: 248-479-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...