1-(2,6-Diethylphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

018555-5G
FLUO018555-5GEA 149 EUR
FLUO018555-5G
1-(2,6-Diethylphenyl)-2-thioharnstoff
1-(2,6-Diethylphenyl)-2-thioharnstoff
Formel: (C₆H₃(CH₂CH₃)₂)NC(S)NH₂
Molecular Weight: 208,33 g/mol
Siedepunkt: 336 °C (1013 hPa)
Schmelzpunkt: 196 °C
MDL: MFCD00041166
CAS-Nummer: 25343-30-0
UN: 2811
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR