3,4-Difluortoluol
Lieferant: FLUOROCHEM
018556-5G
FLUO018556-5GEA
41.4
EUR
FLUO018556-5G
3,4-Difluortoluol
3,4-Difluortoluol
Formel:
C₇H₆F₂ Siedepunkt: 110…113 °C (1013 hPa) Dichte: 1,12 g/cm³ (20 °C) |
MDL:
MFCD00075087 CAS-Nummer: 2927-34-6 UN: 2388 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...