3,4-Dihydroxybenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 3,4-Dihydroxybenzohydrazid, 3,4-Dihydroxybenzol-1-carbohydrazid, 3,4-Dihydroxybenzoesäurehydrazid

018559-10G
FLUO018559-10GEA 269 EUR
FLUO018559-10G
3,4-Dihydroxybenzhydrazid
3,4-Dihydroxybenzhydrazid
Formel: C₇H₈N₂O₃
Molecular Weight: 168.15 g/mol
MDL: MFCD00017064
CAS-Nummer: 39635-11-5
EINECS: 254-550-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR