3,5-Dihydroxybenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 3,5-Dihydroxybenzoic hydrazide

018560-10G
FLUO018560-10GEA 128 EUR
FLUO018560-10G
3,5-Dihydroxybenzhydrazid
3,5-Dihydroxybenzhydrazid
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
MDL: MFCD00017066
CAS-Nummer: 7732-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR