2,6-Diisopropylphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1,3-Diisopropyl-2-isothiocyanatobenzene

018561-10G
FLUO018561-10GEA 329 EUR
FLUO018561-10G
2,6-Diisopropylphenylisothiocyanat
2,6-Diisopropylphenylisothiocyanat
Formel: C₁₃H₁₇NS
Molecular Weight: 219,35 g/mol
Siedepunkt: 84 °C (0,2 mmHg)
MDL: MFCD00041341
CAS-Nummer: 25343-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR