2',4'-Dimethoxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2,4-Dimethoxyphenyl)ethan-1-on

018567-25G
FLUO018567-25GEA 79.1 EUR
FLUO018567-25G
2',4'-Dimethoxyacetophenon
2',4'-Dimethoxyacetophenon
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
MDL: MFCD00008727
CAS-Nummer: 829-20-9
EINECS: 212-587-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR