Veratrohydrazid

Lieferant: FLUOROCHEM

Synonyme: 3,4-Dimethoxybenzoesäurehydrazid, 3,4-Dimethoxybenzol-1-carbohydrazid, 3,4-Dimethoxybenzohydrazid, 3,4-Dimethoxybenzhydrazid

018570-10G
FLUO018570-10GEA 0 EUR
FLUO018570-10G
Veratrohydrazid
Veratrohydrazid
Formel: C₉H₁₂N₂O₃
Molecular Weight: 196,21 g/mol
MDL: MFCD00017061
CAS-Nummer: 41764-74-3
EINECS: 255-541-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR