2,3-Dimethoxybenzylchlorid

Lieferant: FLUOROCHEM

018573-5G
FLUO018573-5GEA 0 EUR
FLUO018573-5G
2,3-Dimethoxybenzylchlorid
2,3-Dimethoxybenzylchlorid
Formel: C₉H₁₁ClO₂
Molecular Weight: 186,64 g/mol
MDL: MFCD01310812
CAS-Nummer: 3893-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR