2,5-Dimethoxybenzylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-(Chloromethyl)-1,4-dimethoxybenzene

018574-5G
FLUO018574-5GEA 225 EUR
FLUO018574-5G
2,5-Dimethoxybenzylchlorid
2,5-Dimethoxybenzylchlorid
Formel: C₉H₁₁ClO₂
Molecular Weight: 186,64 g/mol
MDL: MFCD00092195
CAS-Nummer: 3840-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR