3,4-Dimethoxyzimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3,4-Dimethoxyphenyl)propensäure, 3,4-Dimethoxycinnamsäure

018578-10G
FLUO018578-10GEA 0 EUR
FLUO018578-10G
3,4-Dimethoxyzimtsäure
3,4-Dimethoxyzimtsäure
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,21 g/mol
Siedepunkt: 343 °C (1013 hPa)
Schmelzpunkt: 198…202 °C
MDL: MFCD00004387
CAS-Nummer: 2316-26-9
EINECS: 219-025-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR