(3,5-Dimethoxy-4-hydroxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid

018581-1G
FLUO018581-1GEA 146 EUR
FLUO018581-1G
(3,5-Dimethoxy-4-hydroxyphenyl)essigsäure
(3,5-Dimethoxy-4-hydroxyphenyl)essigsäure
Formel: C₁₀H₁₂O₅
Molecular Weight: 212,2 g/mol
MDL: MFCD00016834
CAS-Nummer: 4385-56-2
EINECS: 224-494-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR