1,3-Dimethoxy-4-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 1-Iod-2,4-dimethoxybenzol

018584-10G
FLUO018584-10GEA 171 EUR
FLUO018584-10G
1,3-Dimethoxy-4-iodbenzol
1,3-Dimethoxy-4-iodbenzol
Formel: C₈H₉IO₂
Molecular Weight: 264,06 g/mol
MDL: MFCD00059268
CAS-Nummer: 20469-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR