3,4-Dimethoxy-β-methyl-β-nitrostyrol

Lieferant: FLUOROCHEM
Achtung

018586-10G
FLUO018586-10GEA 0 EUR
FLUO018586-10G
3,4-Dimethoxy-β-methyl-β-nitrostyrol
3,4-Dimethoxy-β-methyl-β-nitrostyrol
Formel: C₁₁H₁₃NO₄
Molecular Weight: 223.23 g/mol
MDL: MFCD00024815
CAS-Nummer: 122-47-4
EINECS: 204-547-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR