4,5-Dimethoxy-2-nitrobenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: 6-Nitroveratrumylalkohol, 6-Nitroveratrylalkohol, (4,5-Dimethoxy-2-nitrophenyl)methanol

018587-1G
FLUO018587-1GEA 31.1 EUR
FLUO018587-1G
4,5-Dimethoxy-2-nitrobenzylalkohol
4,5-Dimethoxy-2-nitrobenzylalkohol
Formel: C₉H₁₁NO₅
MDL: MFCD00014701
CAS-Nummer: 1016-58-6
EINECS: 213-806-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR