3,4-Dimethoxy-β-nitrostyrol

Lieferant: FLUOROCHEM

Synonyme: 1,2-Dimethoxy-4-(2-nitrovinyl)benzene

018588-10G
FLUO018588-10GEA 0 EUR
FLUO018588-10G
3,4-Dimethoxy-β-nitrostyrol
3,4-Dimethoxy-β-nitrostyrol
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.19 g/mol
MDL: MFCD00024823
CAS-Nummer: 4230-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR