4,5-Dimethoxy-2-nitrotoluol

Lieferant: FLUOROCHEM

018589-5G
FLUO018589-5GEA 372 EUR
FLUO018589-5G
4,5-Dimethoxy-2-nitrotoluol
4,5-Dimethoxy-2-nitrotoluol
Formel: C₉H₁₁NO₄
MDL: MFCD00060708
CAS-Nummer: 7509-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR