(2,3-Dimethoxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(2,3-Dimethoxyphenyl)acetic acid

018593-1G
FLUO018593-1GEA 168 EUR
FLUO018593-1G
(2,3-Dimethoxyphenyl)essigsäure
(2,3-Dimethoxyphenyl)essigsäure
Formel: (C₆H₃(OCH₃)₂)CH₂COOH
Molecular Weight: 196,2 g/mol
MDL: MFCD00060351
CAS-Nummer: 90-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR