(2,5-Dimethoxyphenyl)essigsäure

Lieferant: FLUOROCHEM

018594-5G
FLUO018594-5GEA 125 EUR
FLUO018594-5G
(2,5-Dimethoxyphenyl)essigsäure
(2,5-Dimethoxyphenyl)essigsäure
Formel: (C₆H₃(OCH₃)₂)CH₂COOH
Molecular Weight: 196,2 g/mol
Siedepunkt: 327 °C (1013 hPa)
Schmelzpunkt: 123 °C
MDL: MFCD00004322
CAS-Nummer: 1758-25-4
EINECS: 217-151-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR