(3,5-Dimethoxyphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 3,5-Dimethoxybenzylcyanid

018595-1G
FLUO018595-1GEA 55.2 EUR
FLUO018595-1G
(3,5-Dimethoxyphenyl)acetonitril
(3,5-Dimethoxyphenyl)acetonitril
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.2
Siedepunkt: 298
Schmelzpunkt: 53
MDL: MFCD00016395
CAS-Nummer: 13388-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR