3-(2,5-Dimethoxyphenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(2,5-Dimethoxyphenyl)propanoic acid

018596-1G
FLUO018596-1GEA 71 EUR
FLUO018596-1G
3-(2,5-Dimethoxyphenyl)propionsäure
3-(2,5-Dimethoxyphenyl)propionsäure
Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
MDL: MFCD00060325
CAS-Nummer: 10538-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR