1-(2,4-Dimethoxyphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dimethoxyphenylthiourea, N-(2,4-Dimethoxyphenyl)thiourea

018599-10G
FLUO018599-10GEA 279 EUR
FLUO018599-10G
1-(2,4-Dimethoxyphenyl)-2-thioharnstoff
1-(2,4-Dimethoxyphenyl)-2-thioharnstoff
Formel: (C₆H₃(OCH₃)₂)NC(S)NH₂
Molecular Weight: 212,27 g/mol
Siedepunkt: 337 °C (1013 hPa)
Schmelzpunkt: 194 °C
MDL: MFCD00041158
CAS-Nummer: 35696-77-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR