1-(2,5-Dimethoxyphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: 2,5-Dimethoxyphenylthiourea, N-(2,5-Dimethoxyphenyl)thiourea

018600-10G
FLUO018600-10GEA 318 EUR
FLUO018600-10G
1-(2,5-Dimethoxyphenyl)-2-thioharnstoff
1-(2,5-Dimethoxyphenyl)-2-thioharnstoff
Formel: (C₆H₃(OCH₃)₂)NC(S)NH₂
Molecular Weight: 212,27 g/mol
Siedepunkt: 337 °C (1013 hPa)
Schmelzpunkt: 163 °C
MDL: MFCD00022163
CAS-Nummer: 67617-98-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR