4-(Dimethylamino)benzhydrazid
Lieferant: FLUOROCHEM
018603-10G
FLUO018603-10GEA
131
EUR
FLUO018603-10G
4-(Dimethylamino)benzhydrazid
4-(Dimethylamino)benzhydrazid
Formel:
C₉H₁₃N₃O |
MDL:
MFCD00014761 CAS-Nummer: 19353-92-5 EINECS: 242-985-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...