4-(Dimethylamino)zimtsäure

Lieferant: FLUOROCHEM

Synonyme: 4-(Dimethylamino)cinnamic Acid

018604-5G
FLUO018604-5GEA 74.7 EUR
FLUO018604-5G
4-(Dimethylamino)zimtsäure
4-(Dimethylamino)zimtsäure
Formel: C₁₁H₁₃NO₂
MDL: MFCD00004397
CAS-Nummer: 1552-96-1
EINECS: 216-299-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR