Bis(4-methylphenyl)methanol

Lieferant: FLUOROCHEM

Synonyme: 4,4'-Dimethyldiphenylmethanol, 4,4'-Dimethylbenzhydrylalkohol, Bis(4-methylphenyl)methanol, 4,4'-Dimethyl-α-phenylbenzylalkohol, Bis(4-methylphenyl)carbinol

018608-5G
FLUO018608-5GEA 393 EUR
FLUO018608-5G
Bis(4-methylphenyl)methanol
4,4'-Dimethylbenzhydrol
Formel: C₁₅H₁₆O
Molecular Weight: 212,29 g/mol
MDL: MFCD00017216
CAS-Nummer: 885-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR