2,3-Dimethylbenzonitril

Lieferant: FLUOROCHEM

018609-10G
FLUO018609-10GEA 94.1 EUR
FLUO018609-10G
2,3-Dimethylbenzonitril
2,3-Dimethylbenzonitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
MDL: MFCD00019744
CAS-Nummer: 5724-56-1
EINECS: 227-229-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR