2,4-Dimethylbenzonitril

Lieferant: FLUOROCHEM

018610-10G
FLUO018610-10GEA 323 EUR
FLUO018610-10G
2,4-Dimethylbenzonitril
2,4-Dimethylbenzonitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
MDL: MFCD00016377
CAS-Nummer: 21789-36-6
EINECS: 244-580-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR