2,6-Dimethylbenzonitril

Lieferant: FLUOROCHEM

018612-5G
FLUO018612-5GEA 48.8 EUR
FLUO018612-5G
2,6-Dimethylbenzonitril
2,6-Dimethylbenzonitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
MDL: MFCD00037166
CAS-Nummer: 6575-13-9
EINECS: 229-503-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR