2,3-Dimethylbenzylamin

Lieferant: FLUOROCHEM

018613-2G
FLUO018613-2GEA 0 EUR
FLUO018613-2G
2,3-Dimethylbenzylamin
2,3-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135.21 g/mol
MDL: MFCD00052680
CAS-Nummer: 51586-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR