2,4-Dimethylbenzylamin

Lieferant: FLUOROCHEM

018614-5G
FLUO018614-5GEA 306 EUR
FLUO018614-5G
2,4-Dimethylbenzylamin
2,4-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
MDL: MFCD00025575
CAS-Nummer: 94-98-4
EINECS: 202-380-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR