2,5-Dimethylbenzylamin

Lieferant: FLUOROCHEM

Synonyme: (2,5-Dimethylphenyl)methanamine

018615-1G
FLUO018615-1GEA 45.9 EUR
FLUO018615-1G
2,5-Dimethylbenzylamin
2,5-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
MDL: MFCD00039780
CAS-Nummer: 93-48-1
EINECS: 202-250-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR