3,4-Dimethylbenzylamin

Lieferant: FLUOROCHEM

018617-5G
FLUO018617-5GEA 148 EUR
FLUO018617-5G
3,4-Dimethylbenzylamin
3,4-Dimethylbenzylamin
Formel: C₉H₁₃N
MDL: MFCD00025577
CAS-Nummer: 102-48-7
EINECS: 203-034-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR