4-Chlormethyl-m-xylol

Lieferant: FLUOROCHEM

018618-10G
FLUO018618-10GEA 203 EUR
FLUO018618-10G
4-Chlormethyl-m-xylol
4-Chlormethyl-m-xylol
Formel: C₉H₁₁Cl
Molecular Weight: 154,64 g/mol
Siedepunkt: 30…36 °C (0,2 mmHg)
Dichte: 1,058 g/cm³ (19 °C)
MDL: MFCD00015556
CAS-Nummer: 824-55-5
EINECS: 212-531-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR