4-Chlormethyl-o-xylol

Lieferant: FLUOROCHEM

Synonyme: 3,4-Dimethylbenzylchlorid, 1-(Chlormethyl)-3,4-dimethylbenzol, α-Chlor-1,3,4-trimethylbenzol

018620-10G
FLUO018620-10GEA 53 EUR
FLUO018620-10G
4-Chlormethyl-o-xylol
4-Chlormethyl-o-xylol
Formel: C₉H₁₁Cl
Molecular Weight: 154,64 g/mol
MDL: MFCD00000910
CAS-Nummer: 102-46-5
EINECS: 203-032-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR