Dimethyl-5-bromisophthalat
Lieferant: FLUOROCHEM
Synonyme:
5-Brom-1,3-benzoldicarbonsäuredimethylester, 5-Bromisophthalsäuredimethylester
018622-10G
FLUO018622-10GEA
0
EUR
FLUO018622-10G
Dimethyl-5-bromisophthalat
Dimethyl-5-bromisophthalat
Formel:
C₁₀H₉BrO₄ Molecular Weight: 273,08 g/mol |
MDL:
MFCD00078709 CAS-Nummer: 51760-21-5 EINECS: 257-386-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...