1-(2,4-Dimethylphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
Synonyme:
1-(2,4-Xylyl)thioharnstoff, N-(2,4-Dimethylphenyl)thioharnstoff, 2,4-Dimethylphenylthioharnstoff
018638-10G
FLUO018638-10GEA
141
EUR
FLUO018638-10G
1-(2,4-Dimethylphenyl)-2-thioharnstoff
1-(2,4-Dimethylphenyl)-2-thioharnstoff
Formel:
(C₆H₃(CH₃)₂)NC(S)NH₂ Molecular Weight: 180,27 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 188…190 °C |
MDL:
MFCD00041164 CAS-Nummer: 16738-20-8 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...