1-(2,6-Dimethylphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
2,6-Dimethylphenylthioharnstoff, 1-(2,6-Xylyl)thioharnstoff, N-(2,6-Dimethylphenyl)thioharnstoff
018639-5G
FLUO018639-5GEA
167
EUR
FLUO018639-5G
1-(2,6-Dimethylphenyl)-2-thioharnstoff
1-(2,6-Dimethylphenyl)-2-thioharnstoff
Formel:
(C₆H₃(CH₃)₂)NC(S)NH₂ Molecular Weight: 180,27 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 210 °C |
MDL:
MFCD00041165 CAS-Nummer: 6396-76-5 EINECS: 229-005-8 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...