1-(2,4-Dimethylphenyl)-2-harnstoff

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dimethylphenylurea

018640-5G
FLUO018640-5GEA 137 EUR
FLUO018640-5G
1-(2,4-Dimethylphenyl)-2-harnstoff
1-(2,4-Dimethylphenyl)-2-harnstoff
Formel: C₉H₁₂N₂O
Molecular Weight: 164,20 g/mol
MDL: MFCD00025409
CAS-Nummer: 2990-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR