(3,5-Dinitro-4-hydroxyphenyl)essigsäure
Lieferant: FLUOROCHEM
018643-1G
FLUO018643-1GEA
31.1
EUR
FLUO018643-1G
(3,5-Dinitro-4-hydroxyphenyl)essigsäure
(3,5-Dinitro-4-hydroxyphenyl)essigsäure
Formel:
C₈H₆N₂O₇ Molecular Weight: 242,14 g/mol |
MDL:
MFCD00016995 CAS-Nummer: 10463-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...