1,3-Diphenyl-2-thioharnstoff
Lieferant: FLUOROCHEM
Synonyme:
N,N'-Diphenylthioharnstoff, Thiocarbanilid
018646-25G
FLUO018646-25GEA
50.3
EUR
FLUO018646-25G
1,3-Diphenyl-2-thioharnstoff
1,3-Diphenyl-2-thioharnstoff
Formel:
C₁₃H₁₂N₂S Molecular Weight: 228,32 g/mol Siedepunkt: 365 °C (1013 hPa) Schmelzpunkt: 150…153 °C Dichte: 1,3205 g/cm³ (20 °C) |
MDL:
MFCD00004921 CAS-Nummer: 102-08-9 EINECS: 203-004-2 UN: 2811 ADR: 6.1,II Merck Index: 13,03366 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...