1,3-Di(p-tolyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

018648-10G
FLUO018648-10GEA 222 EUR
FLUO018648-10G
1,3-Di(p-tolyl)-2-thioharnstoff
1,3-Di(p-tolyl)-2-thioharnstoff
Formel: C₁₅H₁₆N₂S
MDL: MFCD00008541
CAS-Nummer: 621-01-2
EINECS: 210-665-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR