4'-Ethoxyacetophenon
Lieferant: FLUOROCHEM
018653-10G
FLUO018653-10GEA
0
EUR
FLUO018653-10G
4'-Ethoxyacetophenon
4'-Ethoxyacetophenon
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164.2 g/mol Siedepunkt: 83 °C (0.04 mmHg) Schmelzpunkt: 39 |
MDL:
MFCD00009095 CAS-Nummer: 1676-63-7 EINECS: 216-825-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...