2-Ethoxybenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 2-Ethoxybenzol-1-carbohydrazid, 2-Ethoxybenzohydrazid, 2-Ethoxybenzoesäurehydrazid

018657-5G
FLUO018657-5GEA 64.8 EUR
FLUO018657-5G
2-Ethoxybenzhydrazid
2-Ethoxybenzhydrazid
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180.21 g/mol
MDL: MFCD00017050
CAS-Nummer: 21018-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR