3-Ethoxybenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
3-Ethoxybenzoic hydrazide
018658-5G
FLUO018658-5GEA
414
EUR
FLUO018658-5G
3-Ethoxybenzhydrazid
3-Ethoxybenzhydrazid
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180.21 g/mol |
MDL:
MFCD00017063 CAS-Nummer: 27830-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...