4-Ethoxybenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 4-Ethoxybenzoic hydrazide

018659-10G
FLUO018659-10GEA 279 EUR
FLUO018659-10G
4-Ethoxybenzhydrazid
4-Ethoxybenzhydrazid
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180.21 g/mol
MDL: MFCD00017074
CAS-Nummer: 58586-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR