3-Ethoxybenzonitril

Lieferant: FLUOROCHEM

018662-2G
FLUO018662-2GEA 0 EUR
FLUO018662-2G
3-Ethoxybenzonitril
3-Ethoxybenzonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00001805
CAS-Nummer: 25117-75-3
EINECS: 246-632-2
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR