(2-Ethoxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Ethoxyphenylacetic acid, 2-(2-Ethoxyphenyl)acetic acid

018675-1G
FLUO018675-1GEA 60.9 EUR
FLUO018675-1G
(2-Ethoxyphenyl)essigsäure
(2-Ethoxyphenyl)essigsäure
Formel: H₃CCH₂O(C₆H₄)CH₂COOH
Molecular Weight: 180,2 g/mol
MDL: MFCD00016824
CAS-Nummer: 70289-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR