α-Chlor-4-ethyltoluol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: α-Chlor-p-ethyltoluol

018680-10G
FLUO018680-10GEA 308 EUR
FLUO018680-10G
α-Chlor-4-ethyltoluol
α-Chlor-4-ethyltoluol
Formel: C₉H₁₁Cl
Molecular Weight: 154,64 g/mol
MDL: MFCD00039357
CAS-Nummer: 1467-05-6
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR