Ethyl-1-naphthoat
Lieferant: FLUOROCHEM
018686-10G
FLUO018686-10GEA
58.1
EUR
FLUO018686-10G
Ethyl-1-naphthoat
Ethyl-1-naphthoat
Formel:
C₁₃H₁₂O₂ Molecular Weight: 200,24 g/mol |
MDL:
MFCD00016805 CAS-Nummer: 3007-97-4 EINECS: 221-120-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...